Learn More
R&D Systems™ Recombinant Human Ubiquitin N-Terminal Biotin Protein
Description
Specifications
Specifications
| Accession Number | P0CG47.1 |
| Formulation | Lyophilized from a solution in HEPES. |
| Gene ID (Entrez) | 7314 |
| Name | Ubiquitin N-Terminal Biotin |
| Quantity | 50μg |
| Source | E. coli-derived human Ubiquitin protein Gln2 - Gly76 with a single N-terminal biotin |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Conjugate | Biotin |
| Recombinant | Recombinant |
| Purity or Quality Grade | >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.