Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human VEGF 165 Protein, Animal-Free

Catalog No. p-7250171
Click to view available options
Quantity:
1 mg
50 μg
This item is not returnable. View return policy
This item is not returnable. View return policy
Measured in a cell proliferation assay using HUVEC human umbilical vein endothelial cells. Conn, G. et al. (1990) Proc. Natl. Acad. Sci. USA 87:1323. The ED50 for this effect is 1.50-12.0 ng/mL.The specific activity of recombinant human VEGF165 is >8.0 x 105 units/mg, which is calibrated against the human VEGF165 WHO standard (NIBSC code: 02/286).

Specifications

Accession Number NP_001165097.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Sodium Acetate.
Gene ID (Entrez) 7422
Molecular Weight (g/mol) M.W.-observed: 19-21 kDa, under reducing conditions., M.W.-theroretical: 19.2 kDa (monomer)
Quantity 1 mg
Source E. coli-derived human VEGF protein Ala27-Arg191 Produced using non-animal reagents in an animal-free laboratory.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Alias MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in sterile deionized water.
Purity or Quality Grade >97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein VEGF
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.