Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human VEGF-B 167 Protein

Catalog No. 751VEB025
Change view
Click to view available options
25 ug
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
75-1VE-B025 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 75-1VE-B025 Supplier R&D Systems™ Supplier No. 751VEB025
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measured by its binding ability in a functional ELISA.

When Recombinant Rat Neuropilin-1 Fc Chimera is immobilized at 1 μg/mL (100 μL/well), Recombinant Human VEGF-B 167 binds with an ED50 of 10 to 80 ng/mL.

Specifications

Accession Number AAA91463
For Use With (Application) Bioactivity
Gene ID (Entrez) 7423
Molecular Weight (g/mol) 19 kDa (monomer)
Name vascular endothelial growth factor B, VEGFB, VEGF-B, VEGF-related factor, VRFVEGFL
Quantity 25 μg
Source E. coli-derived human VEGF-B protein Pro22-Arg188
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 250 μg/mL in 4mM HCl.
Buffer Lyophilized from a 0.2 μm filtered solution in HCl with Trehalose.
Endotoxin Level <0.10 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.