Learn More
Creative Biomart Recombinant Human VEGFA protein, His-tag, GMP Grade
Supplier:  Creative Biomart VEGFA332THP
Encompass_Preferred
Recombinant Human VEGFA protein GMP Grade(NP_001165097.1)(193 aa) was expressed in HEK293 cells with a C-terminal His Tag.
Catalog No. 50-208-5246