Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human VLDLR Protein

Catalog No. 8444VL025
Change view
Click to view available options
25ug; Carrier Free
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
84-44V-L025 25ug; Carrier Free
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 84-44V-L025 Supplier R&D Systems™ Supplier No. 8444VL025
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human VLDL R Protein is derived from HEK293. The Recombinant Human VLDL R Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P98155
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 7436
Molecular Weight (g/mol) 86kDa
Name VLDLR
Preparation Method Reconstitute at 250μg/mL in sterile PBS.
Quality Control Testing SDS-PAGE: 116-142kDa, reducing conditions
Quantity 25μg
Source Human embryonic kidney cell,HEK293-derived human VLDL R protein Thr25-Ser797,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Termination Thr25
Without Additives Carrier Protein
Gene Alias CARMQ1, CHRMQ1, FLJ35024, very low density lipoprotein receptor, very low-density lipoprotein receptor, VLDL R, VLDL receptor, VLDL-R, VLDLRCH
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE with silver staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.