Learn More
Biomatik Corporation Recombinant Human Von Hippel-Lindau disease tumor suppressor (VHL)
Supplier:  Biomatik Corporation RPC22221
Encompass_Preferred
Recombinant Human Von Hippel-Lindau disease tumor suppressor (VHL) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: VHL. Target Synonyms: Protein G7pVHL. Accession Number: P40337. Expression Region: 1~213aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 28.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7943