Learn More
Creative Biomart GMP Grade Recombinant Human VTN protein
Supplier:  Creative Biomart VTN599THP
Encompass_Preferred
Recombinant Human VTN protein GMP Grade(NP_000629.3)(465 aa) was expressed in HEK293 cells with a C-terminal His Tag.
Catalog No. 50-208-5267