Learn More
R&D Systems™ Recombinant Human Wnt/RSPO3 Agonist Protein, CF
Description
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Molecular Weight (g/mol) | 83-98 kDa, under reducing conditions. |
| Quantity | 1 mg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human Wnt/RSPO3 protein Proprietary |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1μg of the protein by the LAL method. |
| Gene Alias | CRISTIN1, Protein with TSP type-1 repeat, PWTSR, roof plate-specific spondin-3, RSPO 3, R-spondin 3 homolog, R-spondin-3, Thrombospondin type-1 domain-containing protein 2, THSD2, Wnt |
| Conjugate | Unconjugated |
| Species | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.