Learn More
Proteintech Group Inc Recombinant Human Wnt3A 100ug
SDP
Supplier:  Proteintech Group Inc HZ1296100UG
Encompass_Preferred
Recombinant Human Wnt3A is a member of the Wnt gene family that consists of structurally linked genes that encode secreted signaling proteins. These proteins are associated with oncogenesis and in numerous developmental processes. These processes include regulation of cell fate and patterning during embryogenesis. Human Wnt3A is a protein which shows 84% identity relative to other human Wnt3 proteins. Wnt3A plays a key role in maintaining the integrity of embryonic and adult tissues. The post-translational glycosylation and acylation of Wnts are essential for their efficient secretion and biological activity.
Catalog No. 50-197-3966