Learn More
Abcam Recombinant Human Wnt5a protein, 25UG
Supplier:  Abcam AB20462725UG
Encompass_Preferred
Recombinant Human Wnt5a protein is a Human Full Length protein, in the 62 to 380 aa range, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: Protein Wnt-5a, WNT5A
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2150603