Learn More
Active Motif Recombinant IKKβ protein
SDP
Supplier:  Active Motif 81066
Encompass
Recombinant human IKKb protein was expressed in a baculovirus expression system as the full-length protein (accession number NP_001547.1) with an N-terminal FLAG tag. The molecular weight of the protein is 87.8 kDa.
Catalog No. NC1851399