Learn More
Biomatik Corporation Recombinant Influenza A virus Nucleoprotein (NP)
Supplier:  Biomatik Corporation RPC22678
Encompass_Preferred
Recombinant Influenza A virus Nucleoprotein (NP) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Influenza A virus (strain A/Puerto Rico/8/1934 H1N1). Target Name: NP. Target Synonyms: NP; Nucleoprotein; Nucleocapsid protein; Protein N. Accession Number: P03466. Expression Region: 1~498aa. Tag Info: N-Terminal 6Xhis-B2M-Tagged. Theoretical MW: 70.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7940