Learn More
R&D Systems™ Recombinant Influenza A Virus H1N1 Hemagglutinin Protein
Description
Specifications
Specifications
| Accession Number | AAD17229.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS with Trehalose. |
| Molecular Weight (g/mol) | M.W.-observed: 55-70 kDa, under reducing conditions., M.W.-theroretical: 58 kDa |
| Quantity | 100 μg |
| Source | Spodoptera frugiperda, Sf 21 (baculovirus)-derived influenza a virus h1n1 Hemagglutinin protein Asp18-Gln529, with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Purity or Quality Grade | >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.