Learn More
Biomatik Corporation Recombinant Influenza B virus Nucleoprotein (NP)
Supplier:  Biomatik Corporation RPC22762
Encompass_Preferred
Recombinant Influenza B virus Nucleoprotein (NP) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Influenza B virus (strain B/Lee/1940). Target Name: NP. Target Synonyms: Nucleocapsid protein; Protein N. Accession Number: P04665. Expression Region: 1~560aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 77.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7939