Learn More
Creative Biomart Peginterferon alfa-2a
Supplier:  Creative Biomart THP0022
Encompass_Preferred
The product is a form of recombinant interferon and is derived from the alfa-2a moeity of recombinant human interferon and acts by binding to human type 1 interferon receptors.
Catalog No. 50-208-4073