Learn More
Abcam Recombinant Macaca fascicularis Claudin18.2 protein - Active
Catalog No. 502699188
Encompass_Preferred
Recombinant Macaca fascicularis Claudin18.2 protein - Active is a Full Length protein, in the 1 to 261 aa range, expressed in Mammalian, < 1 EU/µg endotoxin level and suitable for WB, FuncS.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-269-9188 Supplier Abcam Supplier No. AB31453720UG
Recombinant Macaca fascicularis Claudin18.2 protein - Active is a Full Length protein, in the 1 to 261 aa range, expressed in Mammalian, < 1 EU/µg endotoxin level and suitable for WB, FuncS.
The product is subject to the following: Abcam Restricted Use Statement