Learn More
Abcam Recombinant Macaca fascicularis ENPP3/B10 protein - Active
Catalog No. 502699190
Encompass_Preferred
Recombinant Macaca fascicularis ENPP3/B10 protein - Active is a Fragment protein, in the 46 to 874 aa range, expressed in Mammalian, with >95% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE, FuncS.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-269-9190 Supplier Abcam Supplier No. AB31453620UG
Recombinant Macaca fascicularis ENPP3/B10 protein - Active is a Fragment protein, in the 46 to 874 aa range, expressed in Mammalian, with >95% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE, FuncS.
The product is subject to the following: Abcam Restricted Use Statement