Learn More
Active Motif Recombinant MAX protein
SDP
Supplier:  Active Motif 81017
Encompass
Recombinant MAX protein was expressed in a baculovirus expression system as the full-length protein (accession number NP_660087.1) with an N-terminal FLAG tag. The molecular weight of MAX protein is 18.5 kDa.
Catalog No. NC1851007