Learn More
R&D Systems™ Recombinant MERS-CoV 3CL Protease
Description
Specifications
Specifications
| Accession Number | K9N638 |
| For Use With (Application) | Bioactivity |
| Formulation | 1.7 mg/ml (50 μM) in 50 mM HEPES pH 7.5, 150 mM NaCl, 1 mM EDTA, 1 mM DTT |
| Gene ID (Entrez) | 43740578 |
| Molecular Weight (g/mol) | M.W.-theoretical: 33 kDa |
| Quantity | 50 μg |
| Source | E. coli-derived mers-cov 3 CL Protease protein Ser3248 - Gln3553 Contains the following substitutions: Val 3551 to Leu; Met 3552 to Phe |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 48 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Gene Alias | 3 CL Proteinase, 3 C-like main protease, 3 CL-Mpro, COVID-19, M Proteinase, Main Protease, Mpro |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.