Learn More
Active Motif Recombinant Mononucleosomes (H3.1) - biotinylated
SDP
Supplier:  Active Motif 31467
Encompass
Recombinant Nucleosomes (H3.1) - biotinylated, Human consists of 5000 bp of DNA (plasmid) and two molecules each of histones H2A that includes amino acids 1-130 (end) (accession number NM_003512), H2B that includes amino acids 1-126 (end) (accession number NM_003518), H3.1 that includes amino acids 1-136 (end) (accession number NM_003529) with a C-terminal Biotin tag, and H4 that includes amino acids 1-103 (end) (accession number NM_003548). Recombinant Nucleosomes (H3.1) - biotinylated, Human was generated in E. coli cells and has an observed molecular weight of 110 kDa. Nucleosomes comprise the smallest subunit of chromatin; they consist of 147 bp of DNA wrapped around an octamer of core histone proteins (H2A, H2B, H3 and H4). Therefore, nucleosomes […] For more information, please visit: https://www.activemotif.com/catalog/details/31467.
Catalog No. NC1846033