Learn More
Biomatik Corporation Recombinant Mouse Acyl-CoA-binding protein (Dbi)
Supplier:  Biomatik Corporation RPC26167
Encompass_Preferred
Recombinant Mouse Acyl-CoA-binding protein (Dbi) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Dbi. Target Synonyms: Diazepam-binding inhibitor (DBI; Endozepine; EP; ACBP). Accession Number: P31786. Expression Region: 2~87aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 16.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4635