Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Aminopeptidase PILS/ARTS1 Protein

Catalog No. 2500ZN010
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2500ZN010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 2500ZN010 Supplier R&D Systems™ Supplier No. 2500ZN010
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Measured by its ability to cleave the fluorogenic peptide substrate, Leu-AMC. The specific activity is >100 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number Q6GTP5
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl and Glycerol.
Gene ID (Entrez) 51752
Molecular Weight (g/mol) M.W. (Observed): 111 kDa, reducing conditions; M.W. (theoretical): 105 kDa
Name Aminopeptidase PILS/ARTS1
Quantity 10 μg
Source Mouse myeloma cell line, NS0-derived mouse Aminopeptidase PILS/ARTS1 protein Ser27-Leu930, with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol Erap1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Aminopeptidase PILS/ARTS1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.