Learn More
R&D Systems™ Recombinant Mouse ASAH2 Protein
Description
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in MES, NaCl and Sodium Cholate. |
| Gene ID (Entrez) | 54447 |
| Molecular Weight (g/mol) | 81 kDa |
| Name | ASAH2/N-acylsphingosine Amidohydrolase-2 |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse ASAH2/N-acylsphingosine Amidohydrolase-2 protein Thr34-Thr756,with an N-terminal 6-His tag Accession &Num; NP_061300 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.