Learn More
Biomatik Corporation Recombinant Mouse B-lymphocyte antigen CD20 (Ms4a1), partial
Supplier:  Biomatik Corporation RPC24831
Encompass_Preferred
Recombinant Mouse B-lymphocyte antigen CD20 (Ms4a1), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Ms4a1. Target Synonyms: B-cell differentiation antigen Ly-44Lymphocyte antigen 44Membrane-spanning 4-domains subfamily A member 1; CD20. Accession Number: P19437. Expression Region: 111~291aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 22.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6277