Learn More
R&D Systems™ Recombinant Mouse BTNL10/Butyrophilin-like 10 Fc Protein
Description
Specifications
Specifications
| Accession Number | Q9JK39 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 100129094 |
| Molecular Weight (g/mol) | M.W. (Observed): 58-63 kDa, reducing conditions; M.W. (theoretical): 42.6 kDa |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line, NS0-derived mouse BTNL10 protein Mouse BTNL10 (Lys30-Glu167) (N-terminus) Accession # Q9JK39 IEGRMDP Mouse IgG2a (Glu98-Lys330) (C-terminus) |
| Storage Requirements | 12 months from date of receipt, <= -20 °C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution. |
| Regulatory Status | RUO |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.