Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse BTNL2/Butyrophilin-like 2 Protein

Catalog No. 8605BT050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8605BT050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 8605BT050 Supplier R&D Systems™ Supplier No. 8605BT050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Learn More About B7-related Butryophilins as Immune Modulators View Webinar

Measured by its ability to inhibit IL-2 secretion by mouse T cells in the presence of anti-CD3. The ED50 for this effect is 1-6 μg/mL.

Specifications

Accession Number O70355
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 56244
Molecular Weight (g/mol) M.W. (Observed): 54-67 kDa, reducing conditions; M.W. (theoretical): 49 kDa
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived mouse BTNL2/Butyrophilin-like 2 protein Asp27-Ser452, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias BTL-II, Butyrophilin like 2, Butyrophilin-like 2, butyrophilin-like 2 (MHC class II associated)
Common Name BTNL2/Butyrophilin-like 2
Gene Symbol Btnl2
Biological Activity Measured by its ability to inhibit IL-2 secretion by mouse T cells in the presence of anti-CD3. The ED50 for this effect is typically 1-6μg/mL.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Reconstitution Reconstitute at 200 μg/mL in PBS.
Expression System Mouse myeloma cell line, NS0-derived Asp27-Ser452, with a C-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE with silver staining.
Protein BTNL2/Butyrophilin-like 2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.