Learn More
R&D Systems™ Recombinant Mouse Carbonic Anhydrase IX Protein
Description
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in Tris and NaCl. |
| Gene ID (Entrez) | 230099 |
| Molecular Weight (g/mol) | 40 kDa |
| Name | Carbonic Anhydrase IX/CA9 |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse Carbonic Anhydrase IX/CA9 protein Gln32-Asp390,with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.