Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Cathepsin A/Lysosom Carboxypeptidase A

Catalog No. 9789SE020
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
9789SE020 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 9789SE020 Supplier R&D Systems™ Supplier No. 9789SE020
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse Cathepsin A/Lysosom Carboxypeptidase A is derived from NS0. The Recombinant Mouse Cathepsin A/Lysosom Carboxypeptidase A has been validated for the following applications: Enzyme Activity.

Specifications

Accession Number P16675
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 5476
Molecular Weight (g/mol) 53 kDa
Name beta-galactosidase 2, beta-galactosidase protective protein, Carboxypeptidase C, Carboxypeptidase L, cathepsin ANGBE, CTSA, EC 3.4.16, EC 3.4.16.5, GLB2, GSL, Lysosomal Carboxypeptidase A, lysosomal protective protein, PPCA, PPGBprotective protein for beta-galactosidase (galactosialidosis), Protective protein cathepsin A, Protective protein for beta-galactosidase
Quantity 20 μg
Source Mouse myeloma cell line,NS0-derived mouse Cathepsin A/Lysosomal Carboxypeptidase A protein Ala24-Tyr474,with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Conjugate Unconjugated
Species Mouse
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.