Learn More
R&D Systems™ Recombinant Mouse CD19 Fc Chimera Protein
Description
Specifications
Specifications
| Accession Number | P25918 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 930 |
| Molecular Weight (g/mol) | 57 kDa |
| Name | B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, Leu-12, MGC12802, T-cell surface antigen Leu-12 |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived mouse CD19 protein, (N-terminus) Mouse CD19 (Arg19-Gly287) Accession &Num; P25918 IEGRMDP Mouse IgG2a (Glu98-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <1.0 EU per 1μg of the protein by the LAL method. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.