Learn More
R&D Systems™ Recombinant Mouse CD200R1L Fc Chimera Protein
Description
Specifications
Specifications
| Accession Number | Q6XJV6.1 |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 344807 |
| Molecular Weight (g/mol) | Observed: 65-85 kDa, under reducing conditions; Theroretical: 48 kDa |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line, NS0-derived mouse CD200R1L protein Mouse CD200R1L (Ile25-Thr220) ) (N-terminus) Accession # Q6XJV6.1 IEGRMDP Mouse IgG2a (Glu98-Lys330 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Purity or Quality Grade | >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.