Learn More
R&D Systems™ Recombinant Mouse CEACAM-5/CD66e Fc Chimera Protein
Description
Specifications
Specifications
| Accession Number | Q3UKK2.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 1048 |
| Molecular Weight (g/mol) | M.W.-observed: 125-145 kDa and 185-195 kDa, under reducing conditions, M.W.-theoretical: 130 kDa |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line, NS0-derived mouse CEACAM-5/CD66e protein Mouse CEACAM-5/CD66e (Gln35-Glu947) (N-terminus) Accession # Q3UKK2.1 IEGRMDP Mouse IgG2a (Glu98-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e, CD66e antigen, CEA, CEACAM5, CEACD66e, DKFZp781M2392, Meconium antigen 100 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.