Learn More
R&D Systems™ Recombinant Mouse Coagulation Factor XIV/Protein C

Description
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in Sodium Acetate and NaCl. |
| Gene ID (Entrez) | 19123 |
| Molecular Weight (g/mol) | 49 kDa (Pro) & 48 kDa (Mature) |
| Name | Coagulation Factor XIV/Protein C |
| Quantity | 10 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived mouse Coagulation Factor XIV/Protein C protein Ile19-Leu460 (pro) & Ala42-Leu460 (mature),both with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Structural Form | Pro and Mature forms |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.