Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Endoglycan/PODXL2 Protein

Catalog No. 8365EG050
Change view
Click to view available options
50ug; Carrier Free
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
83-65E-G050 50ug; Carrier Free
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 83-65E-G050 Supplier R&D Systems™ Supplier No. 8365EG050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse Endoglycan/PODXL2 Protein is derived from NS0. The Recombinant Mouse Endoglycan/PODXL2 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number XP_006506309
Formulation Lyophilized from a 0.2μm filtered solution in PBS
Gene ID (Entrez) 319655
Molecular Weight (g/mol) 51kDa
Name Endoglycan/PODXL2
Preparation Method Reconstitute at 100μg/mL in PBS.
Quality Control Testing SDS-PAGE: 90-124kDa, reducing conditions
Quantity 50μg
Source Mouse myeloma cell line,NS0-derived mouse Endoglycan/PODXL2 protein Gly30-Gly498,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Termination Gly30
Without Additives Carrier Protein
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE with silver staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.