Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse EpCAM/TROP1 Fc Chimera Protein

Catalog No. 9364EP100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
93-64E-P100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 93-64E-P100 Supplier R&D Systems™ Supplier No. 9364EP100
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse EpCAM/TROP1 Fc Chimera Protein is derived from NS0. The Recombinant Mouse EpCAM/TROP1 Fc Chimera Protein has been validated for the following applications: Bioactivity.

Order Info

Ships at ambient temperature.

Specifications

Accession Number Q99JW5
Formulation Lyophilized from a 0.2μm filtered solution in PBS
Gene ID (Entrez) 4072
Molecular Weight (g/mol) 55 kDa
Quantity 100 μg
Source Mouse myeloma cell line,NS0-derived mouse EpCAM/TROP1 protein, (N-terminus) Mouse EpCAM/TROP1 (Gln24 & Arg81-Thr266) Accession &Num; Q99JW5 IEGRMDP Mouse IgG2a (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method
Biological Activity 0.4 to 2.4μg/mL
Product Type Recombinant protein
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.