Learn More
Biomatik Corporation Recombinant Mouse Erythroferrone (Erfe)
Supplier:  Biomatik Corporation RPC25432
Encompass_Preferred
Recombinant Mouse Erythroferrone (Erfe) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Erfe. Target Synonyms: Complement C1q tumor necrosis factor-related protein 15Myonectin. Accession Number: Q6PGN1. Expression Region: 25~340aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 35.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4928