Learn More
R&D Systems™ Recombinant Mouse Fc gamma RIII/CD16 Protein
Description
Specifications
Specifications
| Accession Number | P08508.1 |
| Formulation | Supplied as a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 14131 |
| Molecular Weight (g/mol) | 30-45 kDa, reducing conditions |
| Name | Fc gamma RIII (CD16) |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse Fc gamma RIII (CD16) protein Ala31-Thr215,with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening. 3 months, -20 to -70° C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.