Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse FGF-17 Protein

Catalog No. p-4721476
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in MOPS, (NH 4 ) 2 SO 4 DTT and EDTA with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in MOPS, (NH 4 ) 2 SO 4 DTT and EDTA.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
7400FG025 Lyophilized from a 0.2 μm filtered solution in MOPS, (NH 4 ) 2 SO 4 DTT and EDTA with BSA as a carrier protein.
7400FG025CF Lyophilized from a 0.2 μm filtered solution in MOPS, (NH 4 ) 2 SO 4 DTT and EDTA.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 7400FG025 Supplier R&D Systems™ Supplier No. 7400FG025
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse FGF-17 Protein is derived from E. coli. The Recombinant Mouse FGF-17 Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2 μm filtered solution in MOPS, (NH 4 ) 2 SO 4 DTT and EDTA with BSA as a carrier protein.
Gene ID (Entrez) 14171
Molecular Weight (g/mol) 22.7 kDa
Name FGF-17
Quantity 25 μg
Source E. coli-derived mouse FGF-17 protein Thr23-Thr216,with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.