Learn More
Abcam Recombinant mouse FGF2 protein (Active), 10UG
Supplier:  Abcam AB28546510UG
Encompass_Preferred
Recombinant mouse FGF2 protein (Active) is a Mouse Full Length protein, expressed in Escherichia coli, with >=95% purity and < 1 EU/µg endotoxin level.
Protein/ Peptide/ Enzyme Synonyms: Fgf-2, Fgf2, Fibroblast growth factor 2, FGF-2, Basic fibroblast growth factor, Heparin-binding growth factor 2, bFGF, HBGF-2
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-221-5549