Learn More
Abcam Recombinant Mouse Fibroblast activation protein, alpha (Tagged), 25UG
Supplier:  Abcam AB27150625UG
Encompass_Preferred
Recombinant Mouse Fibroblast activation protein, alpha (Tagged) is a Mouse Fragment protein, in the 26 to 761 aa range, expressed in HEK 293, with =90% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: Prolyl endopeptidase FAP, Dipeptidyl peptidase FAP, Fibroblast activation protein alpha, Gelatine degradation protease FAP, Integral membrane serine protease, Post-proline cleaving enzyme, Serine integral membrane protease, Surface-expressed protease, FAPalpha, SIMP, Seprase, Fap
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1898091