Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Fibulin 7 Protein

Catalog No. 8606FB050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8606FB050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 8606FB050 Supplier R&D Systems™ Supplier No. 8606FB050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse Fibulin 7 Protein is derived from CHO. The Recombinant Mouse Fibulin 7 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q501P1
For Use With (Application) Bioactivity
Gene ID (Entrez) 129804
Molecular Weight (g/mol) Observed M.W.: 53-63kDa, reducing conditions, Theoretical M.W.: 47kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line,CHO-derived mouse Fibulin 7 protein Gln25-Phe440,with an N-terminal HA Tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias FBLN7, FIBL-7, fibulin 7, TM14
Common Name TM14
Gene Symbol Fbln7
Biological Activity Measured by the ability of the immobilized protein to support the adhesion of SVEC4-10 mouse vascular endothelial cells. The ED50 for this effect is typically 0.7-3.5μg/mL.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Gln25-Phe440, with an N-terminal HA Tag
Purity or Quality Grade >90%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.