Learn More
R&D Systems™ Recombinant Mouse gAdiponectin/gAcrp30 Protein
Description
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 11450 |
| Molecular Weight (g/mol) | Observed M.W.: 18-24 kDa, reducing conditions; Theoretical M.W.: 16.7 kDa (monomer) |
| Name | gAdiponectin/gAcrp30 |
| Quantity | 25 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse gAdiponectin/gAcrp30 protein Ala111-Asn247 & Tyr112-Asn247,both with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening. 3 months, -20 to -70° C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.