Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Galectin-1 Protein
SDP

Catalog No. 1245GA050CF
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and DTT with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and DTT.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
1245GA050CF Lyophilized from a 0.2μm filtered solution in PBS, EDTA and DTT.
1245GA050 Lyophilized from a 0.2μm filtered solution in PBS, EDTA and DTT with BSA as a carrier protein.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 1245GA050CF Supplier R&D Systems™ Supplier No. 1245GA050/CF

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse Galectin-1 Protein is derived from E. coli. The Recombinant Mouse Galectin-1 Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS, EDTA and DTT.
Gene ID (Entrez) 16852
Molecular Weight (g/mol) 15 kDa
Name Galectin-1
Quantity 50 μg
Source E. coli-derived mouse Galectin-1 protein Ala2-Glu135
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Product Line R&D Systems
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.