Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse GDF-3 Protein

Catalog No. 9009GD010CF
Change view
Click to view available options
:
10μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
90-09G-D010CF 10μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 90-09G-D010CF Supplier R&D Systems™ Supplier No. 9009GD010/CF
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse GDF-3 Protein is derived from CHO. The Recombinant Mouse GDF-3 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q07104
For Use With (Application) Bioactivity
Gene ID (Entrez) 9573
Molecular Weight (g/mol) Observed M.W.: 14-23kDa, reducing conditions, Theoretical M.W.: 13kDa
Quantity 10μg
Source Chinese Hamster Ovary cell line,CHO-derived mouse GDF-3 protein Ala253-Gly366
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias GDF3, GDF-3, growth differentiation factor 3, growth/differentiation factor 3, KFS3, MCOP7, MCOPCB6, Vgr-2
Common Name GDF-3
Gene Symbol Gdf3
Biological Activity Measured by its ability to induce Smad2 phosphorylation in P19 mouse embryonal carcinoma cells. Mazerbourg, S. et al. (2004) Mol. Endocrinol. 18:653. 50-100ng/mL of Recombinant Mouse GDF-3 can effectively induce Smad2 phosphorylation.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Ala253-Gly366
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.