Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse GDF-8 Propeptide Fc Chimera Protein

Catalog No. 6986PG025CF
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in MES, NaCl and CHAPS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in MES, NaCl and CHAPS.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
6986PG025CF Lyophilized from a 0.2μm filtered solution in MES, NaCl and CHAPS.
6986PG025 Lyophilized from a 0.2μm filtered solution in MES, NaCl and CHAPS with BSA as a carrier protein.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 6986PG025CF Supplier R&D Systems™ Supplier No. 6986PG025/CF
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse GDF-8 Propeptide Fc Chimera Protein is derived from NS0. The Recombinant Mouse GDF-8 Propeptide Fc Chimera Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in MES, NaCl and CHAPS.
Gene ID (Entrez) 17700
Molecular Weight (g/mol) 54.9 kDa (monomer)
Name GDF-8/Myostatin Propeptide Fc Chimera
Quantity 25 μg
Source Mouse myeloma cell line,NS0-derived mouse GDF-8/Myostatin protein, (N-terminus) Mouse GDF-8 Propeptide (Asn25-Arg267) with a substitution Ser265Arg Accession &Num; NP_034964 IEGRMDP Mouse IgG2A (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Gene Alias GDF8, GDF-8, GDF8growth differentiation factor 8, growth/differentiation factor 8, MSLHP, MSTN, Myostatin
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.