Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse GITR/TNFRSF18 Fc Chimera Protein

Catalog No. 524GR050
Change view
Click to view available options
50ug; Carrier Free
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
524GR050 50ug; Carrier Free
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 524GR050 Supplier R&D Systems™ Supplier No. 524GR050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. Immobilized rmGITR/Fc Chimera at 2 μg/mL (100 μL/well) can bind rmGITR Ligand with a linear range of 0.8-50 ng/mL.

Specifications

Accession Number Q8C4K3
For Use With (Application) Binding Activity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 8784
Molecular Weight (g/mol) M.W. (Observed): 55-65 kDa, reducing conditions; M.W. (theoretical): 42 kDa (monomer)
Name GITR/TNFRSF18 Fc Chimera
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived mouse GITR/TNFRSF18 protein Mouse GITR (Ser22-His153) (N-terminus) Accession # Q8C4K3 IEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol Tnfrsf18
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein GITR/TNFRSF18
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.