Learn More
Abcam Recombinant Mouse IgG2a Fc (Active) protein, 500UG
Catalog No. 502703458
Encompass_Preferred
Recombinant Mouse IgG2a Fc (Active) protein is a Mouse Fragment protein, in the 98 to 330 aa range, expressed in HEK 293, with >95% purity and < 1 EU/µg endotoxin level.
Protein/ Peptide/ Enzyme Synonyms: Immunoglobulin heavy chain gamma polypeptide, Ighg
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-3458 Supplier Abcam Supplier No. AB316059500UG
Recombinant Mouse IgG2a Fc (Active) protein is a Mouse Fragment protein, in the 98 to 330 aa range, expressed in HEK 293, with >95% purity and < 1 EU/µg endotoxin level.
Protein/ Peptide/ Enzyme Synonyms: Immunoglobulin heavy chain gamma polypeptide, Ighg
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement