Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse IL-11 Protein, CF

Catalog No. 11585IL100
Change view
Click to view available options
Quantity:
100 μg
20 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-585-IL100 100 μg
11-585-IL050 50 μg
11-585-IL020 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 11-585-IL100 Supplier R&D Systems™ Supplier No. 11585IL100
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Measured in a cell proliferation assay using T11 mouse plasmacytoma cells. Nordan, R.P. etáal. (1987) J. Immunol. 139:813. The ED50 for this effect is less than 0.750 ng/mL.

Specifications

Accession Number P47873.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS and EDTA with Trehalose.
Gene ID (Entrez) 3589
Molecular Weight (g/mol) M.W.-observed: 20 kDa, under reducing conditions., M.W.-theroretical: 19
Quantity 100 μg
Source Chinese Hamster Ovary cell line, CHO-derived mouse IL-11 protein Pro22-Leu199
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias Adipogenesis inhibitory factor, AGIF, AGIFoprelvekin, IL11, IL-11Oprelvekin, interleukin 11, interleukin-11, Oprelvekin
Conjugate Unconjugated
Reconstitution Reconstitute the 20μg size at 200μg/mL in PBS. Reconstitute all other sizes at 500μg/mL in PBS.
Protein IL-11
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.