Learn More
Abcam Recombinant mouse IL-15 protein (Active), 50UG
Supplier:  Abcam AB26445650UG
Encompass_Preferred
Recombinant mouse IL-15 protein (Active) is a Mouse Full Length protein, in the 49 to 162 aa range, expressed in Escherichia coli, with >90% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE, FuncS, MS.
Protein/ Peptide/ Enzyme Synonyms: Interleukin-15, IL-15, Il15
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2191140