Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse IL-31 (Mammalian-expressed) Protein

Catalog No. p-7199097
Change view
Click to view available options
Quantity:
10 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-409-ML010 10 μg
10-409-ML050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 10-409-ML010 Supplier R&D Systems™ Supplier No. 10409ML010
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measured by its ability to induce STAT3 reporter activity in HEK293 human embryonic kidney cells transfected with mouse IL-31RA and OSMR beta.

The ED50 for this effect is 0.05-0.4 ng/mL.

Specifications

Accession Number Q6EAL8.1
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 386653
Molecular Weight (g/mol) Observed: 16-25 kDa, under reducing conditions; Theroretical: 15 kDa
Quantity 10 μg
Source Human embryonic kidney cell, HEK293-derived mouse IL-31 protein Ala31-Cys163
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Reconstitution Reconstitute at 100 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Protein IL-31
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.