Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse IL1RAPL1 Fc Chimera Protein

Catalog No. 10756MR050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10756MR050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10756MR050 Supplier R&D Systems™ Supplier No. 10756MR050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Measured by its binding ability in a functional ELISA. When Recombinant Human PTPRD Fc Chimera (Catalog # 9995-PR ) is immobilized at 1 μg/mL (100 μL/well), Recombinant Mouse IL1RAPL1 Fc Chimera (Catalog # 10756-MR) binds with an ED50 of 2.25-18.0 ng/mL.

Specifications

Accession Number P59823.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 11141
Molecular Weight (g/mol) M.W.-observed: 78-88 kDa, under reducing conditions, M.W.-theroretical: 65 kDa
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived mouse IL1RAPL1 protein Mouse IL1RAPL1 (Leu19-Thr357) (N-terminus) Accession # P59823.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias IL1R8IL1RAPL, IL-1R9, IL-1 RAPL1, IL-1-RAPL-1, IL-1 RAPL-1, IL1RAPL-1, interleukin 1 receptor accessory protein-like 1, interleukin 1 receptor-8, interleukin-1 receptor accessory protein-like 1, mental retardation, X-linked 10, MR x 10, MR x 21, MR x 34, oligophrenin-4, OPHN4mental retardation, X-linked 21, Three immunoglobulin domain-containing IL-1 receptor-related 2, TIGIRR-2, TIGIRR-2 mental retardation, X-linked 34, X-linked interleukin-1 receptor accessory protein-like 1
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein IL1RAPL1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.